(±)-3,5,5-Trimethyl-1-hexanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,5,5-Trimethyl-1-hexanol, 3,5,5-Trimethylhexan-1-ol, 3,5,5-Trimethylhexyl alcohol
M-Clarity™ quality level = MQ300
W332402-1KG W332402-20KG W332402-SAMPLE
SIALW332402-1KGEA 86.9 EUR
SIALW332402-1KG SIALW332402-20KG SIALW332402-SAMPLE
(±)-3,5,5-Trimethyl-1-hexanol, Sigma-Aldrich®
(±)-3,5,5-Trimethyl-1-hexanol
Formula: (CH₃)₃CCH₂CH(CH₃)CH₂CH₂OH
Boiling Pt: 193…194 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 80 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00036138
CAS Number: 3452-97-9
EINECS: 222-376-7
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37229e1f94f0ca0beccd95c10e5aa4bc92298d4e&vid=6e796ba4819a3eb496c38b2850887cd5d9dcb820&prid=web&clid=SAPDAM