(±)-1-Octen-3-ol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W280518-1KG-K W280518-20KG-K W280518-4KG-K W280518-8KG-K
SIALW280518-1KG-KEA 180 EUR
SIALW280518-1KG-K SIALW280518-20KG-K SIALW280518-4KG-K SIALW280518-8KG-K
(±)-1-Octen-3-ol, Sigma-Aldrich®
(±)-1-Octen-3-ol
Formula: C₈H₁₆O
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –49 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004589
CAS Number: 3391-86-4
EINECS: 222-226-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=852e188ec9a8f0b95fb8267eecb509cc8b9b4817&vid=db554820fe02a5820107eb97e322b46f3310f665&prid=web&clid=SAPDAM