(±)-3-Quinuclidinol

Supplier: Thermo Scientific

Synonyms: (±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine

BTB12730.1 BTB12730.10 BTB12730.50
MAYBBTB12730.1EA 0 EUR
MAYBBTB12730.1 MAYBBTB12730.10 MAYBBTB12730.50
(±)-3-Quinuclidinol
(±)-3-Quinuclidinol
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8 III
Merck Index: 12,08266

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR