(±)-3-Quinuclidinol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine
M-Clarity™ quality level = MQ100
253340-25G 253340-5G
SIAL253340-25GEA 139 EUR
SIAL253340-25G SIAL253340-5G
(±)-3-Quinuclidinol, Sigma-Aldrich®
(±)-3-Quinuclidinol
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8 III
Merck Index: 12,08266