(±)-1-Phenoxy-2-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Phenoxy-2-propanol, Propylene Phenoxytol, (±)-Monopropylene glycol phenyl ether
M-Clarity™ quality level = MQ100
484423-1L 484423-5L
SIAL484423-1LEA 61.2 EUR
SIAL484423-1L SIAL484423-5L
(±)-1-Phenoxy-2-propanol, Sigma-Aldrich®
(±)-1-Phenoxy-2-propanol
Formula: C₆H₅OCH₂CH(OH)CH₃
MW: 152.19 g/mol
Boiling Pt: 243
Melting Pt: 11.4
Density: 1.0665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016861
CAS Number: 770-35-4
EINECS: 212-222-7