(±)-α-Methylbenzylamine

Supplier: Thermo Scientific
Danger

Synonyms: DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine

SB01486.1 SB01486.10 SB01486.50
MAYBSB01486.1EA 0 EUR
MAYBSB01486.1 MAYBSB01486.10 MAYBSB01486.50
(±)-α-Methylbenzylamine
(±)-α-Methylbenzylamine
Formula: C₆H₅CH(NH₂)CH₃
Boiling Pt: 184…186 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008069
CAS Number: 618-36-0
EINECS: 210-545-8
UN: 2735
ADR: 8,II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7749b7c7d0770d82ae5145b1a236458393427163&vid=9ef2ef0e518b58972043eaec1de61438a07bcc37&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR