(±)-α-Methylbenzylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine
M31104-100ML M31104-25ML M31104-500ML
SIALM31104-100MLEA 82.6 EUR
SIALM31104-100ML SIALM31104-25ML SIALM31104-500ML
(±)-α-Methylbenzylamine, Sigma-Aldrich®
(±)-α-Methylbenzylamine
Formula: C₆H₅CH(NH₂)CH₃
MW: 121.18 g/mol
Boiling Pt: 184…186 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00008069
CAS Number: 618-36-0
EINECS: 210-545-8
UN: 2735
ADR: 8 II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7749b7c7d0770d82ae5145b1a236458393427163&vid=9ef2ef0e518b58972043eaec1de61438a07bcc37&prid=web&clid=SAPDAM