(±)-1-Phenylethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol
M-Clarity™ quality level = MQ100
P13800-1KG P13800-25G P13800-500G
SIALP13800-1KGEA 143 EUR
SIALP13800-1KG SIALP13800-25G SIALP13800-500G
(±)-1-Phenylethanol, Sigma-Aldrich®
(±)-1-Phenylethanol
Formula: C₆H₅CH(OH)CH₃
MW: 122.17 g/mol
Boiling Pt: 401
Melting Pt: 20
Density: 1.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004508
CAS Number: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa955c5829772d9b27ccf96aa5e9b66b661e94a5&vid=43f0c1e8ef3c916809a95af92edf59ec33769eab&prid=web&clid=SAPDAM