(±)-1-Phenyl-1-propanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol

M-Clarity™ quality level = MQ300

W288403-10KG W288403-1KG W288403-5KG W288403-SAMPLE
SIALW288403-10KGEA 1280 EUR
SIALW288403-10KG SIALW288403-1KG SIALW288403-5KG SIALW288403-SAMPLE
(±)-1-Phenyl-1-propanol, Sigma-Aldrich®
(±)-1-Phenyl-1-propanol
Formula: C₆H₅CH(OH)CH₂CH₃
Boiling Pt: ∼219 °C (1013 hPa)
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004564
CAS Number: 93-54-9
EINECS: 202-256-0
Merck Index: 13,03804
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30e80b6acffb0d7728fba7e94ab32f00c90ce567&vid=f2d76406485c12e8edcd1452122e64d6ec3bd99d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR