(±)-3-Carene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3,7,7-Trimethylbicyclo[4.1.0]-3-heptene, 3,7,7-Trimethylbicyclo(4.1.0)hept-3-ene, 3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene, 3-Carene, (±)-3,7,7-Trimethylbicyclo[4.1.0]-3-heptene
M-Clarity™ quality level = MQ100
115576-1L
115576-25ML
SIAL115576-1LEA
141
EUR
SIAL115576-1L
SIAL115576-25ML
(±)-3-Carene, Sigma-Aldrich®
(±)-3-Carene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 167…170 °C (1013 hPa) Density: 0.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001315 CAS Number: 13466-78-9 EINECS: 236-719-3 UN: 2319 ADR: 3 III Merck Index: 12,01885 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...