(±)-3-Bromocyclohexene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
333549-10ML 333549-250ML 333549-50ML
SIAL333549-10MLEA 83.2 EUR
SIAL333549-10ML SIAL333549-250ML SIAL333549-50ML
(±)-3-Bromocyclohexene, Sigma-Aldrich®
(±)-3-Bromocyclohexene
Formula: C₆H₉Br
Boiling Pt: 89…91 °C (13 mmHg)
Density: 1.389 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00013775
CAS Number: 1521-51-3