Glutaraldehyde 25% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
M-Clarity™ quality level = MQ300
G5882-100ML G5882-10X10ML G5882-10X1ML G5882-1L G5882-50ML G5882-80ML
SIALG5882-100MLEA 339 EUR
SIALG5882-100ML SIALG5882-10X10ML SIALG5882-10X1ML SIALG5882-1L SIALG5882-50ML SIALG5882-80ML
Glutaraldehyde 25% in aqueous solution, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 188
Melting Pt: –6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1 II
Merck Index: 13,04485