Glutaraldehyde 50% aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
MQ100
49629-1L 49629-2.5L 49629-250ML
SIAL49629-1LEA 94.9 EUR
SIAL49629-1L SIAL49629-2.5L SIAL49629-250ML
Glutaraldehyde 50% aqueous solution, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 188
Melting Pt: -10
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1 II
Merck Index: 13,04485