Glutaraldehyde 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
M-Clarity™ quality level = MQ300
G7651-10ML G7651-10X10ML
SIALG7651-10MLEA 122 EUR
SIALG7651-10ML SIALG7651-10X10ML
Glutaraldehyde 50% in aqueous solution, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1,II