EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM for hardness of water determinations
Supplier: VWR Chemicals
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
28662.290EA 49.1 EUR
28662.290 28662.370
EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM for hardness of water determinations
EDTA disodium salt
1ml = 5.6° dH/100 H₂O
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed