EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution
Supplier: Roth Carl
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
K714.1 K714.2
ROTHK714.1EA 29.4 EUR
ROTHK714.1 ROTHK714.2
EDTA disodium salt 0.1 mol/l (0.2 N) in aqueous solution
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556