Glutaraldehyde 8% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
M-Clarity™ quality level = MQ200
G7526-10ML G7526-10X10ML
SIALG7526-10MLEA 72.3 EUR
SIALG7526-10ML SIALG7526-10X10ML
Glutaraldehyde 8% in aqueous solution, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 188
Melting Pt: -10
Density: 1.014
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1 II
Merck Index: 13,04485