Hydrochloric acid 3.571 mol/l (N/0.28), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Warning
Synonyms: Hydrochloric acid solution, Muriatic acid
5610.1000EA 33.6 EUR
5610.1000 5610.9010
Hydrochloric acid 3.571 mol/l (N/0.28), AVS TITRINORM volumetric solution
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II