Dimethylamine 40% in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
163670010 163670025 163670100
ACRO163670010EA 39.8 EUR
ACRO163670010 ACRO163670025 ACRO163670100
Dimethylamine 40% in aqueous solution
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: 6.9
Melting Pt: –37 °C
Density: 0.89 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3 II
Merck Index: 12,03278
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration with HCl 39.0 to 41.0 %