Dimethylamine 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
426458-100ML 426458-1L 426458-2L
SIAL426458-100MLEA 25.6 EUR
SIAL426458-100ML SIAL426458-1L SIAL426458-2L
Dimethylamine 40% in aqueous solution, Sigma-Aldrich®
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: 6.9
Melting Pt: -92.8
Density: 0.938
Flash Pt: -55
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3 II
Merck Index: 12,03278