Sulphuric acid standardised solution 0.2 N in aqueous solution
Supplier: Thermo Fisher Scientific
45062.M4 45062.K7 45062.K2
45062.M4EA 162 EUR
45062.M4 45062.K7 45062.K2
Sulphuric acid standardised solution 0.2 N in aqueous solution
Sulphuric acid
Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00064589
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II