Methanesulphonic acid 70% ≥70% in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
A17803.AE A17803.AP A17803.0F
A17803.AEEA 27.3 EUR
A17803.AE A17803.AP A17803.0F
Methanesulphonic acid 70% ≥70% in aqueous solution
Methanesulphonic acid
CAS No.: 75-75-2 Molecular Formula: CH3SO3H Formula Weight: 96.10 Flash Point: 110°(230°F) Refractive Index: 1.4150 Merck Reference: 14,5954 MDL No.: MFCD00007518
Formula: CH₃SO₃H
Density: 1.320
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
Merck Index: 14,5954
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM