Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Danger
Synonyms: Potash caustic, Caustic potash
32304.291EA 61.1 EUR
32304.291 32304.427
Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, AVS TITRINORM volumetric solution
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8) II
Merck Index: 12,07806
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
Colouration Max. 100 APHA
NIST traceable Confirmed