Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Danger
Synonyms: Potash caustic, Caustic potash
31308.264EA 30.8 EUR
31308.264 31308.297
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Potassium hydroxide
This product may develop a yellow-brown colour during storage.
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8) II
Merck Index: 12,07806
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed