Hydrochloric acid 25%
Supplier: Roth Carl
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
X897.1 X897.2
ROTHX897.1EA 20.1 EUR
ROTHX897.1 ROTHX897.2
Hydrochloric acid 25%
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II