Silver sulphate 10 g/l sulphuric acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Danger
Synonyms: Disilver sulphate
7147.1000EA 0 EUR
7147.1000 7147.2500
Silver sulphate 10 g/l sulphuric acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 1830
ADR: 8 II
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Suitable for COD determination passes test