Zinc sulphate 2 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
83265-1L-F 83265-250ML-F
SIAL83265-1L-FEA 231 EUR
SIAL83265-1L-F SIAL83265-250ML-F
Zinc sulphate 2 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
Melting Pt: 100
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9 III
Merck Index: 12,10293