Orthophosphoric acid 85% ≥85%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phosphoric acid
M-Clarity™ quality level = MQ200
345245-100ML 345245-500ML
SIAL345245-100MLEA 169 EUR
SIAL345245-100ML SIAL345245-500ML
Orthophosphoric acid 85% ≥85%, Sigma-Aldrich®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344