Orthophosphoric acid 85% ≥85%, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Phosphoric acid
M-Clarity™ quality level = MQ300
W290017-10KG-K
W290017-1KG-K
W290017-25KG-K
SIALW290017-10KG-KEA
155
EUR
SIALW290017-10KG-K
SIALW290017-1KG-K
SIALW290017-25KG-K
Orthophosphoric acid 85% ≥85%, Sigma-Aldrich®
Orthophosphoric acid
Formula:
H₃PO₄ Melting Pt: 28 °C Density: 1.71…1.87 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...