Potassium hydroxide 30% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Potash caustic, Caustic potash
10979.K2 10979.AP
10979.K2EA 86.2 EUR
10979.K2 10979.AP
Potassium hydroxide 30% (w/v) in aqueous solution
Potassium hydroxide
CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Density: 1.28 Appearance: Colourless UN No.: 1814 MDL No.: MFCD00003553
Formula: KOH
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II