Sodium hydroxide 32% in aqueous solution
Supplier: Roth Carl
Danger
T197.1 T197.2 T197.3 T197.5
ROTHT197.1EA 22.6 EUR
ROTHT197.1 ROTHT197.2 ROTHT197.3 ROTHT197.5
Sodium hydroxide 32% in aqueous solution
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II