Sodium hydroxide 50% in aqueous solution
Supplier: Roth Carl
Danger
8655.1 8655.2
ROTH8655.1EA 26.3 EUR
ROTH8655.1 ROTH8655.2
Sodium hydroxide 50% in aqueous solution
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II