Tetraethylammonium hydroxide 20% in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Tetrylammonium hydroxide
L03968.22 L03968.36
L03968.22EA 39.5 EUR
L03968.22 L03968.36
Tetraethylammonium hydroxide 20% in aqueous solution
Tetraethylammonium hydroxide
CAS No.: 77-98-5 Molecular Formula: C8H21NO Formula Weight: 147.26 Refractive Index: 1.3730 Beilstein Reference: 3912961 Merck Reference: 14,9201 MDL No.: MFCD00009024
Formula: C₈H₂₁NO
MW: 147.26 g/mol
Density: 1.023
Storage Temperature: Ambient
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 3267
ADR: 8 II
Merck Index: 14,9201