L(+)-Cysteine hydrochloride monohydrate, Sigma-Aldrich®
Supplier: Merck
30129-100G 30129-25G
SIAL30129-100GEA 187 EUR
SIAL30129-100G SIAL30129-25G
L(+)-Cysteine hydrochloride monohydrate, Sigma-Aldrich®
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM