Disodium oxalate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
88626.LQ 88626.LS
88626.LQEA 87.9 EUR
88626.LQ 88626.LS
Disodium oxalate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
di-Sodium oxalate
CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00012465
Formula: Na₂C₂O₄
MW: 134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723