TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Sigma-Aldrich®
Supplier: Merck
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
MQ400
T6666-1KG T6666-250G T6666-50G
SIALT6666-1KGEA 541 EUR
SIALT6666-1KG SIALT6666-250G SIALT6666-50G
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Sigma-Aldrich®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM