Guanidinium hydrochloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
M-Clarity™ quality level = MQ400
G3272-100G
G3272-1KG
G3272-25G
G3272-2KG
G3272-500G
SIALG3272-100GEA
104
EUR
SIALG3272-100G
SIALG3272-1KG
SIALG3272-25G
SIALG3272-2KG
SIALG3272-500G
Guanidinium hydrochloride, Sigma-Aldrich®
Guanidinium hydrochloride
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...