Semicarbazide hydrochloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride
M-Clarity™ quality level = MQ200
S2201-100G S2201-500G S2201-5G
SIALS2201-100GEA 28.8 EUR
SIALS2201-100G SIALS2201-500G SIALS2201-5G
Semicarbazide hydrochloride, Sigma-Aldrich®
Semicarbazide hydrochloride
Formula: CH₆ClN₃O
MW: 111.53 g/mol
Melting Pt: 174…178 °C
MDL Number: MFCD00013009
CAS Number: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,08588