Magnesium acetate tetrahydrate
Supplier: Roth Carl
Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
P026.1 P026.2
ROTHP026.1EA 65.9 EUR
ROTHP026.1 ROTHP026.2
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676