Oxalic acid dihydrate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Ethanedioic acid dihydrate

M-Clarity™ quality level = MQ200

33506-5KG 33506-1KG 33506-250G 33506-25KG 33506-500G 33506-6X1KG
SIAL33506-5KGEA 455 EUR
SIAL33506-5KG SIAL33506-1KG SIAL33506-250G SIAL33506-25KG SIAL33506-500G SIAL33506-6X1KG
Oxalic acid dihydrate, Sigma-Aldrich®
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR