Oxalic acid dihydrate, Sigma-Aldrich®

Supplier: Merck

SIALO0376-500GEA 108 EUR
SIALO0376-500G SIALO0376-10MG SIALO0376-3KG SIALO0376-100G SIALO0376-1KG
Oxalic acid dihydrate, Sigma-Aldrich®
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126,07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101 °C
Density: 1,65 g/cm³ (20 °C)
MDL Number: MFCD00149102
CAS Number: 6153-56-6
UN: 3261
ADR: 8,III
Merck Index: 12,07043
https://ru.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR