EDTA disodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
ED2SS-500G ED2SS-100G ED2SS-1KG ED2SS-2.5KG ED2SS-250G ED2SS-50G ED2SS-5KG
SIALED2SS-500GEA 133 EUR
SIALED2SS-500G SIALED2SS-100G SIALED2SS-1KG SIALED2SS-2.5KG SIALED2SS-250G SIALED2SS-50G SIALED2SS-5KG
EDTA disodium salt dihydrate, Sigma-Aldrich®
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM