EDTA tetrasodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tetrasodium ethylenediaminetetraacetate dihydrate, Edetic acid tetrasodium salt dihydrate, Kestranal® 4S 2 H2O, Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetraacetic acid sodium hydrate, Tetrasodium Ethylenediaminetetraacetate Dihydrate, Kestranal® 4S dihydrate
M-Clarity™ quality level = MQ200
E5391-20KG E5391-250G
SIALE5391-20KGEA 4810 EUR
SIALE5391-20KG SIALE5391-250G
EDTA tetrasodium salt dihydrate, Sigma-Aldrich®
EDTA tetrasodium salt dihydrate
Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
MW: 416.2 g/mol
Melting Pt: > 300 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150026
CAS Number: 10378-23-1
EINECS: 200-573-9
Merck Index: 13,03544