5-Sulphosalicylic acid dihydrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Hydroxy-5-sulphobenzoic acid dihydrate
M-Clarity™ quality level = MQ200
247006-100G
247006-500G
SIAL247006-100GEA
89.6
EUR
SIAL247006-100G
SIAL247006-500G
5-Sulphosalicylic acid dihydrate, Sigma-Aldrich®
5-Sulphosalicylic acid dihydrate
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O MW: 254.22 g/mol Melting Pt: 224 Density: 1.695 g/cm³ (20 °C) Flash Pt: 150 Storage Temperature: Ambient |
MDL Number:
MFCD00149540 CAS Number: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8 III Merck Index: 13,09054 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...