Dipotassium hydrogen orthophosphate trihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
M-Clarity™ quality level = MQ200
P5504-500G P5504-10KG P5504-1KG P5504-5KG
SIALP5504-500GEA 150 EUR
SIALP5504-500G SIALP5504-10KG SIALP5504-1KG SIALP5504-5KG
Dipotassium hydrogen orthophosphate trihydrate, Sigma-Aldrich®
di-Potassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5