Sodium dihydrogen phosphate monohydrate
Supplier: Roth Carl
Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
K300.1 K300.2 K300.5
ROTHK300.1EA 36 EUR
ROTHK300.1 ROTHK300.2 ROTHK300.5
Sodium dihydrogen phosphate monohydrate
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
Melting Pt: 100 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM