Sodium tungstate dihydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
M-Clarity™ quality level = MQ100
379751-25G 379751-5G
SIAL379751-25GEA 486 EUR
SIAL379751-25G SIAL379751-5G
Sodium tungstate dihydrate, Sigma-Aldrich®
Sodium wolframate dihydrate
Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 100
Density: 3.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847