Aluminium potassium sulphate dodecahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium
M-Clarity™ quality level = MQ200
A7210-100G A7210-500G
SIALA7210-100GEA 109 EUR
SIALA7210-100G SIALA7210-500G
Aluminium potassium sulphate dodecahydrate, Sigma-Aldrich®
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373