Calcium chloride dihydrate
Supplier: Roth Carl
Warning
Synonyms: Calcium Chloride Dihydrate, Calcium dichloride
T885.1 T885.2 T885.3
ROTHT885.1EA 39.1 EUR
ROTHT885.1 ROTHT885.2 ROTHT885.3
Calcium chloride dihydrate
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660