Magnesium chloride hexahydrate
Supplier: Roth Carl
Synonyms: Magnesium dichloride
2189.1 2189.2 2189.5
ROTH2189.1EA 0 EUR
ROTH2189.1 ROTH2189.2 ROTH2189.5
Magnesium chloride hexahydrate
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684